missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher HealthCare™ PROTOCOL™ Color-Coded Prefilled Formalin Containers
Prefilled with 10% neutral buffered formalin
Manufacturer: Fisher HealthCare™ 032059
Description
- Color-coded screw cap with leakproof O-ring
- Autoclavable polypropylene
- Paper labels to indicate date, doctor, patient name or ID, and specimen type
- Graduated
Order Info
The 20/10, 40/20 and 60/30mL containers are packaged in shrink wrapped trays to allow for ease of use, handling and storage.
Specifications
20mL | |
10mL formalin fill | |
Color-coded cap w/leakproof O-ring |
Autoclavable Polypropylene | |
Green | |
Paper |