missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher HealthCare™ PROTOCOL Buffer Solutions

$84.97 - $160.31

Specifications

Boiling Point >100°C
Color Colorless
Format Liquid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity pH Price Quantity  
Catalog Number Mfr. No. Quantity pH Price Quantity  
23-262234 Fisher HealthCare™
262234
32 oz. 6.4 Each for $101.19
Case of 12 Each for $1,214.28
23-262236 Fisher HealthCare™
262236
32 oz. 6.8 Each for $84.97
Case of 12 Each for $1,019.64
23-262235 Fisher HealthCare™
262235
1 gal. 6.4 Each for $160.31
Case of 4 Each for $641.24
23-262237 Fisher HealthCare™
262237
1 gal. 6.8 Each for $148.81
Case of 4 Each for $595.24
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

>100°C
Colorless
Liquid