Fisher Chemical™ pH Indicator Solutions
For use in a variety of pH test sets
Manufacturer:  Fisher Chemical™ SI14500
 View more versions of this product
Catalog No. SI14-500
Specifications Description & Specifications
Specifications
Green
500mL
500mL
3.8 to 5.4
Liquid
16.9 oz.
Bromocresol Green Solution