Please login to your online account to display your discounted pricing

Fisher Chemical™ Harris Modified Method Hematoxylin Stains


For histological and cytological use as a nuclear stain

Manufacturer: fisher chemical™  SH264GH

 View more versions of this product

Catalog No. SH26-4D

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Specifications

Includes Acetic Acid
For Use With (Application) PAP staining
Product Type Harris Modified Method Hematoxylin Stain With Acetic Acid
Quantity 4000mL
pH 2 to 3