Please login to your online account to display your discounted pricing
Fisher Chemical™ Harris Modified Method Hematoxylin Stains
For histological and cytological use as a nuclear stain
Manufacturer: fisher chemical™  SH30500GH
 View more versions of this product
Catalog No. SH30-500D
  • / Each
  • / Case of 6 Each
Description & Specifications
Specifications
Product Type Harris Modified Method Hematoxylin Stain Without Acetic Acid
For Use With (Application) PAP staining
Quantity 500mL