Please login to your online account to display your discounted pricing

Fisher Chemical™ Gill Method Hematoxylin Stains


Manufacturer: fisher chemical™  CS4024GH

 View more versions of this product

Catalog No. CS402-4D

  • / Each
 

Description & Specifications

Specifications

Product Type Gill Method Hematoxylin Stain
Quantity 4 x 4000mL
For Use With (Application) Histological and Cytological Staining
pH 2.1
pH 2.1
Type Gill Method Hematoxylin Stain