Please login to your online account to display your discounted pricing

Fisher Chemical™ Gill Method Hematoxylin Stains


Manufacturer: fisher chemical™  CS4011GH

 View more versions of this product

Catalog No. CS401-1D

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

For Use With (Application) Histological and Cytological Staining
Product Type Gill Method Hematoxylin Stain
Quantity 1000mL
pH 2.1
pH 2.1
Type Gill Method Hematoxylin Stain