missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Chemical™ Gill Method Hematoxylin Stains

$144.44 - $401.48

Specifications

For Use With (Application) Histological and Cytological Staining
pH 2.1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
Catalog Number Mfr. No. Quantity Product Type Price Quantity  
CS401-4D View Documents Fisher Chemical™
CS4014GH
4000mL Gill Method Hematoxylin Stain (Gill-2) Each for $401.48
 
CS400-4D View Documents Fisher Chemical™
CS4004GH
4000mL Gill Method Hematoxylin Stain Each for $282.96
 
CS401-1D View Documents Fisher Chemical™
CS4011GH
1000mL Gill Method Hematoxylin Stain Each for $144.44
Case of 6 Each for $866.64
 
Specifications

Specifications

Histological and Cytological Staining
2.1