Please login to your online account to display your discounted pricing

Fisher BioReagents™ YPD Broth (Powder/Molecular Genetics)


Manufacturer: fisher bioreagents™  BP24692

 View more versions of this product

Catalog No. BP2469-2

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Specifications

Format Powder
Product Type YPD Broth
Packaging Poly bottle
Quantity 2000g
Storage Requirements 4°C
Grade Molecular genetics
Media Type YPD Broth