Please login to your online account to display your discounted pricing

Fisher BioReagents™ YPD Broth (Powder/Molecular Genetics)


Manufacturer: fisher bioreagents™  BP246910

 View more versions of this product

Catalog No. BP2469-10

  • / Each
 

Description & Specifications

Specifications

Format Powder
Quantity 10000g
Packaging Poly bottle
Storage Requirements 4°C
Grade Molecular genetics
Product Type YPD Broth
Media Type YPD Broth