Please login to your online account to display your discounted pricing

Fisher BioReagents™ Yeast Extract (Granulated)


Dried yeast autolysate with a high content of amino nitrogen and water soluble B complex vitamins

Manufacturer: fisher bioreagents™  BP97275

 View more versions of this product

Catalog No. BP97275

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Used for preparing microbiological culture media
Format Dehydrated
Media Type Yeast Extract
Product Type Yeast Extract
Quantity 5kg
Includes Vitamin B complex
Packaging Poly pail