Please login to your online account to display your discounted pricing

Fisher BioReagents™ SurePrep™ Water RNA/DNA Purification Kit


Provides a rapid method for the isolation and purification of total RNA and DNA simultaneously from all microorganisms found in environmental water samples, including bacteria, fungi, and algae

Manufacturer: fisher bioreagents™  BP281425

 View more versions of this product

Catalog No. BP281425

  • / Each
 

Description & Specifications

Specifications

Quantity 25 Preps
Format Spin column, filter column
For Use With (Application) PCR, RT-PCR, QRT-PCR, southern blotting and northern blotting
Includes Lysis solution 15mL, Enzyme incubation Buffer 4mL, Nucleic Acid Elution Buffer 4mL, Nucleic Acid wash solution 11mL, spin columns 25, collection tubes 25, 25 elution tubes, 1.7mL, bead tubes 25, 25 filter column, 0.45μm