Please login to your online account to display your discounted pricing
Fisher BioReagents™ SurePrep™ RNA/DNA/Protein Purification Kit
Rapid method for the isolation and purification of total RNA, genomic DNA and proteins sequentially from a single sample of cultured animal cells, tissue samples, blood, bacteria, yeast, fungi or plants
Manufacturer: fisher bioreagents™  BP28024
 View more versions of this product
Catalog No. BP28024
  • / Each
Description & Specifications
Specifications
Includes Lysis solution 40mL, RNA wash solution 20mL, RNA elution solution 6mL, genomic DNA wash solution 15mL, genomic DNA Elution Buffer 15mL, Protein column regeneration Buffer 30mL, Protein column activation and Wash Buffer 60mL, Protein pH Binding Buffer 4mL, Protein Elution Buffer 8mL, Protein neutralizer 2mL, Protein loading dye 2mL, spin columns 50, collection tubes 200, product inserts 1
For Use With (Application) PCR, RT-PCR, reverse transcription PCR, northern blotting, RNase protection, expression array assays, DNA sequencing, southern blotting, SNP analysis, SDS-PAGE and western Blots
Yield 15μg RNA HeLa cells (1 x 106 cells), 8μg DNA HeLa cells (1 x 106 cells), 150μg Protein HeLa cells (1 x 106 cells)
Format Spin column
Quantity 4 Preps
Quantity 4 Preps
Format Spin column