Please login to your online account to display your discounted pricing

Fisher BioReagents™ SurePrep™ RNA Cleanup and Concentration Kit

Provides a rapid method to purify and concentrate RNA isolated using other methods such as phenol:chloroform extraction or after enzymatic reactions

$619.22 - $619.22

Specifications

Format Spin column
For Use With (Application) RT-PCR, northern blotting, RNase protection, primer extension and expression array assays
Includes Binding solution 2 x 20mL, wash solution 22mL, RNA Elution Buffer 6mL, collection tubes, spin columns, elution tubes 1.7mL, product insert
View More Specs
Includes:  Binding solution (2 x 20mL), Wash solution (22mL), RNA elution buffer (6mL), Collection tubes (50), Spin columns (50), Elution tubes (1.7mL) (50), Product insert
Products
Catalog Number Mfr. No. Yield Quantity Price Quantity    

BP280950

 
fisher bioreagents™
BP280950
15μg RNA HeLa cells (1 x 106 cells), 8μg DNA HeLa cells (1 x 106 cells), 150μg Protein HeLa cells (1 x 106 cells) 50 Preps Each for $619.22
Description & Specifications

Specifications

Format Spin column
For Use With (Application) RT-PCR, northern blotting, RNase protection, primer extension and expression array assays
Includes Binding solution 2 x 20mL, wash solution 22mL, RNA Elution Buffer 6mL, collection tubes, spin columns, elution tubes 1.7mL, product insert

  • Fast and easy processing using rapid spin-column format
  • Suitable for all sizes of RNA, from large mRNA to microRNA (miRNA)
  • Able to clean up RNA isolated using different methods, including phenol/chloroform extractions
  • Efficient RNA cleanup from various enzymatic reactions such as labeling, DNase treatment and in vitro transcription

Purified RNA is suitable for Quantitative RT-PCR, Northern blotting, RNase protection, primer extension and expression array assays

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.