Please login to your online account to display your discounted pricing

Fisher BioReagents™ Routine DNA Ladder


Ready-to-use DNA size standard is room-temperature stable for up to two years

Manufacturer: fisher bioreagents™  BP2582200

 View more versions of this product

Catalog No. BP2582200

  • / Each
 

Description & Specifications

Specifications

Product Type 1kb Full Scale DNA Ladder
Color Blue
Format Liquid
No. of Bands 16.0
Quantity 1000μL
Color Blue
Reference Band 16
Storage Requirements Room Temperature
Sufficient For 200 Lanes
For Use With (Application) General purpose, wide DNA size range
Format Liquid
Product Name 1kb Full Scale DNA Ladder
For Use With (Application) General purpose, wide DNA size range