Please login to your online account to display your discounted pricing

Fisher BioReagents™ Ribonuclease B (From Bovine Pancreas)


Manufacturer: fisher bioreagents™  BP2678100

 View more versions of this product

Catalog No. BP2678100

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Specifications

Storage Requirements 0°C
Format Solid
Product Name Ribonuclease B
Color White to Off-White
Source Bovine pancreas
Format Solid
Quantity 100mg
Quantity 100mg