missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ Ribonuclease B (From Bovine Pancreas)

$527.80 - $527.80

Specifications

Color White to Off-White
Product Name Ribonuclease B
Source Bovine pancreas
Storage Requirements 0°C
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BP2678100 Fisher BioReagents™
BP2678100
100mg Each for $527.80
Case of 5 Each for $2,639.00
Specifications Description & Specifications

Specifications

White to Off-White
Ribonuclease B
Bovine pancreas
0°C