Please login to your online account to display your discounted pricing

Fisher BioReagents™ RiboLadder™ RNA Standards


For assessment of single-stranded RNA molecules on both native or denaturing agarose and polyacrylamide gels

Manufacturer: fisher bioreagents™  BP281150

 View more versions of this product

Catalog No. BP281150

  • / Each
 

Description & Specifications

Specifications

Storage Requirements −70°C
Product Type 1kb RNA standard
Kit Contents Two vials of RNA standard (25 Dye loads each), 1X loading Buffer that contains Bromophenol Blue which serves as a tracking dye and 2X sample loading Buffer
Quantity 50 Loadings
Color Blue
For Use With (Application) Assessment of single stranded RNA molecules on both native or denaturing Agarose and Polyacrylamide gels
No. of Bands 9
Format Lyophilized