Please login to your online account to display your discounted pricing

Fisher BioReagents™ RiboLadder™ RNA Standards

For assessment of single-stranded RNA molecules on both native or denaturing agarose and polyacrylamide gels

$312.06 - $315.80

Specifications

For Use With (Application) Assessment of single stranded RNA molecules on both native or denaturing Agarose and Polyacrylamide gels
Format Lyophilized
Storage Requirements −70°C
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

BP281050

 
fisher bioreagents™
BP281050
100bp RNA standard Each for $312.06

BP281150

 
fisher bioreagents™
BP281150
1kb RNA standard Each for $315.80
Description & Specifications

Specifications

For Use With (Application) Assessment of single stranded RNA molecules on both native or denaturing Agarose and Polyacrylamide gels
Format Lyophilized
Storage Requirements −70°C

  • Genetically engineered for precise bands
  • Well-spaced bands for easy identification and sizing of unknown RNA fragments and higher intensity reference band for fast size comparison
  • Packaged in two vials (25 loadings each) and lyophilized to decrease thawing-related degradation, to prolong shelf life and to ensure consistent performance
  • Purity level suitable for capillary electrophoresis and bioanalyzer applications
  • Two size ranges suitable for most RNA applications
  • Ideal for ethidium bromide and fluorescent staining