missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ RiboLadder™ RNA 1X Standard Loading Buffer

For use with Riboladder RNA Standards

Supplier:  Fisher BioReagents™ BP2813500

Catalog No. BP2813500



Description

Description

  • Contains bromophenol blue which serves as a tracking dye for Riboladder RNA Standards

Specifications

Specifications

Blue
RNA Sample Loading Buffer
Two vials of RNA standard (25 Dye loads each), 1X loading Buffer that contains Bromophenol Blue which serves as a tracking dye and 2X sample loading Buffer
500 μL
1X
Lyophilized
Assessment of single stranded RNA molecules on both native or denaturing Agarose and Polyacrylamide gels
−70°C
SDS
Documents

Documents

Promotions

Promotions