Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)
Manufacturer:  Fisher BioReagents™ BP1700100
 View more versions of this product
Catalog No. BP1700-100
Specifications Description & Specifications
Specifications
White
-20°C
Proteinase K
100mg
Tritirachium album
Solid
Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases