Please login to your online account to display your discounted pricing

Fisher BioReagents™ PCR Nucleotide Mix


A premixed 10mM solution of dATP, dCTP, dGTP, and dTTP used for PCR

Manufacturer: fisher bioreagents™  BP25651

 View more versions of this product

Catalog No. BP25651

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) PCR
pH 7.5
Purity >98%
Format Liquid
Concentration 10mM
Quantity 1000μL
Storage Requirements -20°C
Color Colorless