Please login to your online account to display your discounted pricing

Fisher BioReagents™ Optizyme™ RNase Inhibitor


Ideal for use in RT-PCR, cDNA synthesis, and in vitro RNA transcription

Manufacturer: fisher bioreagents™  BP32251

 View more versions of this product

Catalog No. BP32251

  • / Each
 

Description & Specifications

Specifications

Product Name OPTIZYME™ RNase Inhibitor
Color Colorless
Format Liquid
For Use With (Application) RT-PCR, cDNA synthesis and in-vitro RNA transcription
Storage Requirements -20°C
Source Porcine
Quantity 2500U
pH 7.6
Concentration 20 to 40U/μL