Please login to your online account to display your discounted pricing

Fisher BioReagents™ OptiGrow™ Preweighed Media

Optimized for superior yield of bacterial culture

$96.82 - $183.55

Specifications

Packaging Pouch
Grade Molecular genetics
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Format Product Type Price Quantity    

BP142610P1

 
fisher bioreagents™
BP142610P1
Culturing of Escherichia coli Solid LB broth, Miller Pack of 10 Each for $96.82

BP142510P1

 
fisher bioreagents™
BP142510P1
Culturing of Escherichia coli Solid LB Agar, Miller Pack of 10 Each for $149.80

BP246810P1

 
fisher bioreagents™
BP246810P1
Used for obtain high yields of plasmid DNA from transformed Escherichia coli Solid Terrific Broth Pack of 10 Each for $183.55
Description & Specifications

Specifications

Packaging Pouch
Grade Molecular genetics

Four classic media, in convenient, preweighed pouches.

  • Preblended growth-promoting components save time and preparation costs
  • Reduce formulation errors
  • Each pouch contains enough material for 1 liter of liquid media
  • No more weighing of dusty powders
  • High quality—assured by stringent testing procedures

  • LB is used for the culturing of E. coli in molecular biology procedures
  • Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E. coli

Supplied as 10 pouches per box.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.