Please login to your online account to display your discounted pricing

Fisher BioReagents™ NZY Broth (Powder/Molecular Genetics)


Manufacturer: fisher bioreagents™  BP246510

 View more versions of this product

Catalog No. BP2465-10

  • / Each
 

Description & Specifications

Specifications

Format Powder
Packaging Poly pail
Quantity 10kg
Product Type NZY Broth
Media Type NZY Broth
Storage Requirements 4°C
Grade Molecular genetics