Please login to your online account to display your discounted pricing

Fisher BioReagents™ NZCYM Broth (Powder/Molecular Genetics)


Manufacturer: fisher bioreagents™  BP2464500

 View more versions of this product

Catalog No. BP2464-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Specifications

Product Type NZCYM Broth
Format Powder
Packaging Poly bottle
Quantity 500g
Media Type NZCYM Broth
Storage Requirements 4°C
Grade Molecular genetics