Please login to your online account to display your discounted pricing

Fisher BioReagents™ NZCYM Broth (Powder/Molecular Genetics)


Manufacturer: fisher bioreagents™  BP24642

 View more versions of this product

Catalog No. BP2464-2

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Specifications

Packaging Poly bottle
Quantity 2000g
Format Powder
Media Type NZCYM Broth
Product Type NZCYM Broth
Storage Requirements 4°C
Grade Molecular genetics