Please login to your online account to display your discounted pricing

Fisher BioReagents™ Nucleotides


Nucleotides for PCR with a purity level of >98% triphosphate

Manufacturer: fisher bioreagents™  BP25624

 View more versions of this product

Catalog No. BP25624

  • / Each
 

Description & Specifications

Specifications

pH 7 to 7.5
Molecular Weight 555.1
Product Type dCTP
Purity >98%
Format Liquid
For Use With (Application) Dideoxy sequencing, mutagenesis and cDNA synthesis
Concentration 100mM
Quantity 400μL
Color Colorless
Product Type dCTP