Please login to your online account to display your discounted pricing

Fisher BioReagents™ Nucleotides


Nucleotides for PCR with a purity level of >98% triphosphate

Manufacturer: fisher bioreagents™  BP25614

 View more versions of this product

Catalog No. BP25614

  • / Each
 

Description & Specifications

Specifications

Product Type dGTP
pH 7 to 7.5
Color Colorless
Concentration 100mM
Format Liquid
Quantity 400μL
Molecular Weight 595.1
Purity >98%
For Use With (Application) Dideoxy sequencing, mutagenesis and cDNA synthesis