Please login to your online account to display your discounted pricing

Fisher BioReagents™ Microbiology Media: LB Broth, Lennox (Powder)


Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

Manufacturer: fisher bioreagents™  BP1427500

 View more versions of this product

Catalog No. BP1427-500

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Preparing bacterial nutrient media
Quantity 500g
Media Format Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
Grade Molecular genetics
Product Type LB broth, Lennox
Packaging Poly bottle
Format Powder
Media Type LB Broth, Lennox

  • Directions for Media Preparation: Suspend 20g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
  • Provides a nutrient-rich environment to enable faster growth of E. coli
  • Format: Dehydrated powder