Please login to your online account to display your discounted pricing

Fisher BioReagents™ Microbiology Media: LB Broth, Lennox (Powder)


Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

Manufacturer: fisher bioreagents™  BP14272

 View more versions of this product

Catalog No. BP1427-2

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Preparing bacterial nutrient media
Description Bacterial Culture
Product Type LB broth, Lennox
Quantity 2000g
Packaging Poly pail
Format Powder
Grade Molecular genetics
Media Type LB Broth, Lennox
Media Format Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L

  • Directions for Media Preparation: Suspend 20g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
  • Provides a nutrient-rich environment to enable faster growth of E. coli
  • Format: Dehydrated powder