Please login to your online account to display your discounted pricing

Fisher BioReagents™ Microbiology Media Additives: Peptone


Additive for preparing bacterial nutrient media

Manufacturer: fisher bioreagents™  BP1420100

 View more versions of this product

Catalog No. BP1420-100

  • / Each
 

Description & Specifications

Specifications

Product Type Peptone
Packaging Poly bottle
Media Type Peptone
Quantity 100g
Format Powder
Grade Molecular genetics
For Use With (Application) Nitrogen in a form readily available to bacteria