missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ Lysozyme, Egg White (White Cryst. Powder)

$164.13 - $1,573.24

Specifications

Color White
Product Type Lysozyme
Format Crystalline Powder
For Use With (Application) Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
Source Egg white
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BP535-1 View Documents Fisher BioReagents™
FLBP5351
1g Each for $164.13
BP535-25 View Documents Fisher BioReagents™
FLBP53525
25g Each for $1,573.24
Case of 6 Each for $9,439.44
BP535-10 View Documents Fisher BioReagents™
FLBP53510
10g Each for $850.42
Case of 6 Each for $5,102.52
BP535-5 View Documents Fisher BioReagents™
FLBP5355
5g Each for $460.29
Description

Description

  • Application: This enzyme is used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
  • Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
  • Enzyme Activity: ≈20,000U/mg dry weight
  • Solubility: Readily soluble at 5mg/mL in 0.1M KH2PO4 to give colorless solution
Specifications

Specifications

White
Crystalline Powder
Egg white
Lysozyme
Used to lyse E. coli for preparation of plasmid DNA from transformed cell cultures
<0°C