Fisher BioReagents™ Lymphocyte Separation Medium-LSM
One-step centrifugation permits separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets
Supplier:  Fisher BioReagents™ BP2663500
Catalog No. BP2663500
Description
Description
  • Formulated for isolation of mononuclear cells from defribrinated or heparinized whole human blood
  • Sterile-filtered solution of a sucrose polymer and diatrizoate salts at a specific gravity of 1.077 to 1.080g/mL at 20°C
Specifications
Specifications
Animal Culture
Sterile
Separation of mononuclear lymphocytes from erythrocytes, polynuclear lymphocytes and most platelets
Lymphocyte separation medium
Poly Bottles
5 x 100 mL