Please login to your online account to display your discounted pricing

Fisher BioReagents™ LB Agar, Miller (Pre-Buffered Capsules)

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

$359.89 - $359.89

Specifications

Format Pre-Buffered Capsules
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Media Type LB Agar
Packaging Poly bottle
Product Type LB Agar, Miller
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

BP9734-500

 
fisher bioreagents™
BP9734500
25 x 20g Each for $359.89
Description & Specifications

Specifications

Format Pre-Buffered Capsules
For Use With (Application) For maintenance and propagation of Escherichia coli in molecular biology procedures
Media Type LB Agar
Packaging Poly bottle
Product Type LB Agar, Miller
Media Format Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L, Agar 15g/L, TRIS/TRIS HCl 1.5g/L

  • Directions for Media Preparation: Add 20 capsules per 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve capsule. Before use, sterilize by autoclaving for 15 minutes.
  • Product is in capsule form
  • Each capsule contains 1g of broth
  • Capsules are prebuffered and set to pH 7.2 at 20°C using a 10mM solution of tris/tris HCl
  • pH will vary with temperature