missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher BioReagents™ LB Agar, Lennox (Granulated)
Used as a solidifying agent to provide the solid medium to support the growth of microbes
Manufacturer: Fisher BioReagents™ BP9745500
Description
- Granulated LB Agar, Lennox media
- CAS: [91079-40-2], [8013-01-2], [7647-14-5], [9002-18-0]
- Store at room temperature
Specifications
Granule | |
Agar | |
Poly Bottle |
Agar | |
Used as a solidifying agent to provide the solid medium to support the growth of microbes | |
500 g |