Please login to your online account to display your discounted pricing
Fisher BioReagents™ TransMax™ Competent Cells
Recommended for Blue/White screening and standard cloning applications
Manufacturer: fisher bioreagents™  BP40053
 View more versions of this product
Catalog No. BP40053
  • / Each
Description & Specifications
Specifications
For Use With (Application) Recommended for Blue/White screening and standard cloning applications, cloning PCR fragments, complex ligations, in-vitro mutagenesis, cDNA and genomic library construction
Efficiency 1 x 109cfu/μg
Formulation IPTG/X-Gal solution
Product Type FB10B
Quantity 20 x 0.05mL
Product Name FB10B
Quantity 20 x 0.05mL