missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ TransMax™ Competent Cells

Recommended for Blue/White screening and standard cloning applications

$309.39 - $1,755.65

Specifications

Formulation IPTG/X-Gal solution
For Use With (Application) Recommended for Blue/White screening and standard cloning applications, cloning PCR fragments, complex ligations, in-vitro mutagenesis, cDNA and genomic library construction
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Efficiency Product Name Quantity Price Quantity  
Catalog Number Mfr. No. Efficiency Product Name Quantity Price Quantity  
BP40001 Fisher BioReagents™
BP40001
1 x 109cfu/μg 5 x 0.2mL Each for $509.41
BP40003 Fisher BioReagents™
BP40003
1 x 109cfu/μg 20 x 0.05mL Each for $839.31
BP40011 Fisher BioReagents™
BP40011
1 x 108cfu/μg 5 x 0.2mL Each for $481.22
BP40031 Fisher BioReagents™
BP40031
1 x 109cfu/μg 5 x 0.2mL Each for $508.80
BP40021 Fisher BioReagents™
BP40021
1 x 107cfu/μg 4 x 0.5mL Each for $309.39
BP40004 Fisher BioReagents™
BP40004
1 x 109cfu/μg 50 x 0.05mL Each for $1,755.65
BP40051 Fisher BioReagents™
BP40051
1 x 109cfu/μg 5 x 0.2mL Each for $650.82
BP40052 Fisher BioReagents™
BP40052
1 x 109cfu/μg 10 x 0.05mL Each for $335.84
BP40053 Fisher BioReagents™
BP40053
1 x 109cfu/μg 20 x 0.05mL Each for $1,034.14
BP400696 Fisher BioReagents™
BP400696
1 x 108cfu/μg 96 x 0.02mL Each for $812.29
BP41001 Fisher BioReagents™
BP41001
1 x 1010cfu/μg 5 x 0.1mL Each for $607.31
Specifications

Description

TransMax competent cells are produced according to rigorous quality control standards to ensure dependable performance and lot-to-lot consistency. Transformations are performed, at least in triplicate, to verify efficiency specifications using pUC19 plasmid DNA.

  • recA1 and endA1 markers minimize recombination and improve plasmid quality and yield
  • Superior quality and consistency; equivalent to widely used host cells such as DH5α and DH10B
  • Offered in a wide range of efficiencies (107 to 1010) and formats (both tubes and 96-well plates)
  • 96-well plates are ideal for high throughput cloning applications

  • Both electrotransformable and chemically competent cells are available

  • Recommended for Blue/White screening and standard cloning applications, cloning PCR fragments, complex ligations, in vitro mutagenesis, cDNA and genomic library construction

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Specifications

IPTG/X-Gal solution
Recommended for Blue/White screening and standard cloning applications, cloning PCR fragments, complex ligations, in-vitro mutagenesis, cDNA and genomic library construction