Please login to your online account to display your discounted pricing

Fisher BioReagents™ Taq DNA Polymerase


Allows up to 100 cycles of amplification without loss of activity

Manufacturer: fisher bioreagents™  FB600020

 View more versions of this product

Catalog No. FB6000-20

  • / Each
 

Description & Specifications

Specifications

Formulation 10X assay buffer A : Tris-HCl 100mM, pH 9 at 25°C, KCl 500mM and MgCl2 15mM solution in a separate vial
Format Liquid
Packaging Poly Tube
Concentration 5U/μL
Storage Buffer Tris-HCl 20mM, pH 7.5 (at room temperature), KCl 100mM, EDTA 0.1mM, DTT 1mM, Glycerol 50%, Tween™ 20 0.5%, Nonidet™ P40™ 0.5%
Product Name Taq DNA Polymerase
Storage Requirements -20°C
For Use With (Application) PCR
Quantity 25 × 100 Units