Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ DNA Ladders


Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze

Manufacturer: fisher bioreagents™  BP2572100

 View more versions of this product

Catalog No. BP2572100

  • / Each
 

Description & Specifications

Specifications

Formulation Premixed with loading dye
Sufficient For 100 Lanes
No. of Bands 8
Color Colorless
Storage Requirements Stable at room temperature for two years.
For Use With (Application) For quick check of PCR or Enzyme digestion
Format Liquid