Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ DNA Ladders


Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze

Manufacturer: fisher bioreagents™  BP2570100

 View more versions of this product

Catalog No. BP2570100

  • / Each
 

Description & Specifications

Specifications

Sufficient For 100 Lanes
Formulation Premixed with loading dye
No. of Bands 14
Color Colorless
Storage Requirements Stable at room temperature for two years.
For Use With (Application) PCR size confirmation
Format Liquid