Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ DNA Ladders


Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze

Manufacturer: fisher bioreagents™  BP2580100

 View more versions of this product

Catalog No. BP2580100

  • / Each
 

Description & Specifications

Specifications

Formulation Premixed with loading dye
Sufficient For 100 Lanes
For Use With (Application) For general purpose, extra large size
No. of Bands 15
Storage Requirements Stable at room temperature for two years.
Format Liquid
Color Colorless