Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ Core Reagent Set B

Optimized for routine PCR applications

Manufacturer: fisher bioreagents™  FB6245

 View all options for this product

Catalog No. FB6245


Description & Specifications


For Use With (Equipment) Fisher BioReagents™ exACTGene™ Complete PCR Kit
Includes Taq DNA Polymerase, 10X Buffer B, MgCl2, dNTPs
Product Type Core Reagent Set B
Quantity 80U
Storage Requirements -20°C

  • Provide an economical alternative for high quality PCR
  • Kit components are also available separately