Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ Control Primers


Optimized for routine PCR applications

Manufacturer: fisher bioreagents™  BP6118

 View more versions of this product

Catalog No. BP6118

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Fisher BioReagents™ exACTGene™ PCR kit
Quantity 1 Kit
Concentration 20μm
Storage Requirements -20°C
Product Type Control Primer No. 3
Volume 100μL