Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ Control Primers


Optimized for routine PCR applications

Manufacturer: fisher bioreagents™  BP6117

 View more versions of this product

Catalog No. BP6117

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
Volume 100μL
For Use With (Equipment) Fisher BioReagents™ exACTGene™ PCR kit
Concentration 20μm
Quantity 1 Kit
Product Type Control Primer No. 2
Concentration 20μm