missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ Control Primers

Optimized for routine PCR applications

$9.29 - $9.78

Specifications

Concentration 20μm
Storage Requirements -20°C
Volume 100μL
View More Specs

Products 3
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
Catalog Number Mfr. No. Product Type Quantity Price Quantity  
BP6118 View Documents Fisher BioReagents™
BP6118
Control Primer No. 3 1 Kit Each for $9.61
 
BP6116 View Documents Fisher BioReagents™
BP6116
Control Primer No. 1 1 Kit Each for $9.78
 
BP6117 View Documents Fisher BioReagents™
BP6117
Control Primer No. 2 1 Kit Each for $9.29
 
Specifications

Specifications

20μm
-20°C
100μL