Please login to your online account to display your discounted pricing

Fisher BioReagents™ exACTGene™ Control Primers


Optimized for routine PCR applications

Manufacturer: fisher bioreagents™  BP6116

 View more versions of this product

Catalog No. BP6116

  • / Each
 

Description & Specifications

Specifications

For Use With (Equipment) Fisher BioReagents™ exACTGene™ PCR kit
Product Type Control Primer No. 1
Quantity 1 Kit
Storage Requirements -20°C
Volume 100μL
Concentration 20μm
Concentration 20μm