Fisher BioReagents™ Hybridization Cocktail, 50% Formamide
Manufacturer:  Fisher BioReagents™ BP157550
Catalog No. BP1575-50
Description
Description
Components: <85.0% Formamide, 2.2% Sodium Citrate Dihydrate, 4.4% Sodium Chloride, and <6.0% Dextran, Hydrogen Sulfate, Sodium Salt.
[75-12-7 (Formamide)]
[6132-04-3 (Sodium Citrate Dihydrate)]
[7647-14-5 (Sodium Chloride)]
[9011-18-1 (Dextran, Hydrogen Sulfate, Sodium Salt)]
Specifications
Specifications
Molecular biology applications
50mL
Formamide <85%, Sodium Citrate Dihydrate 2.2%, Sodium Chloride 4.4% and Dextran <6% , Hydrogen Sulfate, Sodium Salt