Please login to your online account to display your discounted pricing

Fisher BioReagents™ Granulated Agar


Used as a solidifying agent to provide the solid medium to support the growth of microbes

Manufacturer: fisher bioreagents™  BP97442

 View more versions of this product

Catalog No. BP97442

  • / Each
 

Description & Specifications

Specifications

Format Granule
Packaging Poly Pail
Product Type Agar
Quantity 2kg
For Use With (Application) Used as a solidifying agent to provide the solid medium to support the growth of microbes