Please login to your online account to display your discounted pricing

Fisher BioReagents™ EZ-Run™ Protein Gel Solutions


Resolution equal to or better than gradient precast gels

Manufacturer: fisher bioreagents™  BP7715500

 View more versions of this product

Catalog No. BP7715500

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Western blotting, MALDI analysis, Protein sequencing
Formulation 15% protein gel solution
Separation Range 2 to 100dKa
Quantity 500mL